Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm5176 PubMed Links |
Gm5176 - high mobility group box 2 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Phlda1 - pleckstrin homology-like domain, family A, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009344.3 | 1516 | UTR 3 | NP_033370.2 |