Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATAAAGATTTTAACTTGATGAC[G/T]ATCCAATTAAAAATCAATTCATATA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ralgapa1 PubMed Links |
Ralgapa1 - Ral GTPase activating protein, alpha subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003719.2 | Intron | NP_001003719.1 | ||||
NM_001112714.2 | Intron | NP_001106185.1 | ||||
NM_001286263.1 | Intron | NP_001273192.1 | ||||
NM_019994.5 | Intron | NP_064378.4 |