Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCGCGGGGTCCCGCAGGGTTCG[C/T]GCTGGATGTCTCCCGGGCGACCGGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Efcab2 PubMed Links |
Efcab2 - EF-hand calcium binding domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026626.2 | 186 | UTR 5 | NP_080902.1 |