Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Card14 PubMed Links |
Card14 - caspase recruitment domain family, member 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Sgsh - N-sulfoglucosamine sulfohydrolase (sulfamidase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018822.3 | Intron | NP_061292.2 |
Slc26a11 - solute carrier family 26, member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178743.3 | Intron | NP_848858.2 |