Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTGAAGCTATGAATTTGTGAC[A/G]TGTACCACACACAGAGTCTTATGGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Akap6 PubMed Links |
Akap6 - A kinase (PRKA) anchor protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198111.2 | Intron | NP_932779.2 |