Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACACTTTGACTTTTTGTCAAAA[C/T]GGTCAAAACAGTCTCGCAGCCTGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdh4 PubMed Links |
Cdh4 - cadherin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009867.2 | Intron | NP_033997.1 |