Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sstr4 PubMed Links |
Sstr4 - somatostatin receptor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Thbd - thrombomodulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009378.3 | 1583 | Silent Mutation | NP_033404.1 |