Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGGGGCTAAAAAATAAGCATGC[A/G]TCAGTATGGAGCCGCTACCATCTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Syndig1 PubMed Links |
Syndig1 - synapse differentiation inducing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085521.2 | Intron | NP_001078990.1 |