Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTCAGAAATGATTCAAAATGGT[A/G]TATGACAGCTGGCATATTACTTTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nanp PubMed Links |
Nanp - N-acetylneuraminic acid phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026086.2 | Intron | NP_080362.1 |