Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAGAGATCAAGGCCACTTTTTT[A/T]TCTGGACCTTTATTCTTCAGATTGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sgcd PubMed Links |
Sgcd - sarcoglycan, delta (dystrophin-associated glycoprotein) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011891.4 | Intron | NP_036021.1 |