Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGTCCATATAATTGAGGTCAAA[C/T]TGAATTCAAGAAAGTACAACAACTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdh10 PubMed Links |
Cdh10 - cadherin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009865.2 | Intron | NP_033995.1 |