Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAGATGTTTGGTGATCATGGAA[C/T]GTAAGGTTTAATCTTTTGTCAATTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Atp10b PubMed Links |
Atp10b - ATPase, class V, type 10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176999.3 | Intron | NP_795973.2 |