Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTTGCAGGTCACTAAACTAGAA[C/T]TTGGAATGCATTCTTCCATTGAAAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Akap7 PubMed Links |
Akap7 - A kinase (PRKA) anchor protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018747.4 | Intron | NP_061217.3 |