Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCACACTCTTCAATCATTATACT[A/G]AGCTTTTCTGTCTCACCTAGTTTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kpna2 PubMed Links |
Kpna2 - karyopherin (importin) alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010655.3 | 1570 | Silent Mutation | NP_034785.1 |