Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGTTGCTTTCAGGAAACCAGCC[C/T]ACTGTCCTGGAGACAGCAGAAGCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Klhdc8a PubMed Links |
Klhdc8a - kelch domain containing 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144810.5 | 1521 | Silent Mutation | NP_659059.1 |