Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ocstamp PubMed Links |
Ocstamp - osteoclast stimulatory transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029021.1 | 1122 | Silent Mutation | NP_083297.1 |
Slc13a3 - solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Zfp334 - zinc finger protein 334 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |