Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGCGGTCTTCTGACTTTGGGAAG[A/G]CTTGGGGCGTGTACAGATACTTCGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lamb1 PubMed Links |
Lamb1 - laminin B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008482.2 | 746 | Missense Mutation | NP_032508.2 |