Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGATTTCTTCATCTTCAAAATC[A/G]TCAAAGAATAGACCATTTCATAGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lekr1 PubMed Links |
Lekr1 - leucine, glutamate and lysine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037923.4 | Intron | NP_001033012.2 | ||||
NM_001166659.1 | Intron | NP_001160131.1 |