Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGTGGAAAGCTACAAACATGC[C/T]CACCTATGTTTCTGACACTGCATGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rps6ka5 PubMed Links |
Rps6ka5 - ribosomal protein S6 kinase, polypeptide 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153587.2 | Intron | NP_705815.1 |