Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGAAGGTAGCATTACCTGGGG[A/G]ATATAGAAAAGTCCCAGCAGGTTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Casr PubMed Links |
Casr - calcium-sensing receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013803.2 | 1009 | Silent Mutation | NP_038831.2 |