Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCAGCCATGTGTCAGAATGTAC[A/G]CTAGAATAAATGAGGTACTTTAAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ndufs5 PubMed Links |
Ndufs5 - NADH dehydrogenase (ubiquinone) Fe-S protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030274.1 | Intron | NP_001025445.1 |