Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTACAGCAGTGTGCACAGGAAT[C/G]GGGGTGCTGAGGGGGCCGAGAACCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Myocd PubMed Links |
Myocd - myocardin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145136.4 | 972 | Silent Mutation | NP_660118.3 | |||
NM_146386.3 | 972 | Silent Mutation | NP_666498.2 |