Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATAGTCCTAGATCTAGAATGCC[A/G]GCCCCTGACCTCTAGTTGGCCAGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Trmt12 PubMed Links |
Trmt12 - tRNA methyltranferase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026642.2 | 1398 | Missense Mutation | NP_080918.2 |