Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTGACACGCAGGCACTGTCACT[A/G]TGGCAGAGATTCCGGGACTTGAGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rars2 PubMed Links |
Rars2 - arginyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181406.3 | 753 | Silent Mutation | NP_852071.2 |