Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTGGCTTCACCCCTGGCCTGGC[A/G]ATCCTGCAGGTATTGGAGCATCTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mthfd1 PubMed Links |
Mthfd1 - methylenetetrahydrofolate dehydrogenase (NADP+ dependent), methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138745.2 | 389 | Silent Mutation | NP_620084.2 |