Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATAGCTTTGCGTTGCTGCTTGCT[A/G]TGCTACAGGTGAGTGTGGCGCGACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930480K23Rik PubMed Links |
4930480K23Rik - RIKEN cDNA 4930480K23 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Tnfrsf10b - tumor necrosis factor receptor superfamily, member 10b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020275.4 | 312 | Missense Mutation | NP_064671.2 |