Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ckmt1 PubMed Links |
Ckmt1 - creatine kinase, mitochondrial 1, ubiquitous | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Strc - stereocilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080459.2 | 4575 | Silent Mutation | NP_536707.2 |