Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm13610 PubMed Links |
Gm13610 - predicted gene 13610 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Pomt1 - protein-O-mannosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Prrc2b - proline-rich coiled-coil 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159634.1 | 9849 | UTR 3 | NP_001153106.1 | |||
NM_172661.3 | 9849 | UTR 3 | NP_766249.2 |