Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Aif1l PubMed Links |
Aif1l - allograft inflammatory factor 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145144.1 | Intron | NP_660126.1 |
Lamc3 - laminin gamma 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |