Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCCAGAGTGTGTCACACCGGGT[A/G]TGTGACAATCAGACGACGCTGTGAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ostm1 PubMed Links |
Ostm1 - osteopetrosis associated transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172416.3 | 2544 | UTR 3 | NP_766004.1 |