Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAATTGTGCCTAGCCCAAGCCC[A/G]TGTACAGCCCTGTAGAGCTCAGCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cacna2d3 PubMed Links |
Cacna2d3 - calcium channel, voltage-dependent, alpha2/delta subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009785.1 | Intron | NP_033915.1 |