Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCACTAGACATCTCTACCTACT[A/G]GTCTCTCAGAAAAGGGAATGGCCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mb21d2 PubMed Links |
Mb21d2 - Mab-21 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177718.3 | Intron | NP_808386.2 |