Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCCAGGAACCACTTACAGAAGC[C/T]GTTCGGGAGCACCTTTCCCTTGGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Trim55 PubMed Links |
Trim55 - tripartite motif-containing 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081281.1 | 95 | UTR 5 | NP_001074750.1 |