Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGACAGTAGTATCTTAGGCTTA[C/T]TCAACTATGCAAAGTTTCAGATGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ehbp1 PubMed Links |
Ehbp1 - EH domain binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252515.1 | Intron | NP_001239444.1 | ||||
NM_153078.4 | Intron | NP_694718.4 |