Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAACGTCGTCCTCTTTGTTGCTAT[C/T]CACCTTCTGTTGGACGATCTCTAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rap1gds1 PubMed Links |
Rap1gds1 - RAP1, GTP-GDP dissociation stimulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040690.2 | 997 | Missense Mutation | NP_001035780.1 | |||
NM_001286759.1 | 997 | Missense Mutation | NP_001273688.1 | |||
NM_145544.3 | 997 | Missense Mutation | NP_663519.2 |