Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGTCTCTGCTACATAGCAGGAG[G/T]GGGCATGTTAGCCAGAGTGCTCTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fam135b PubMed Links |
Fam135b - family with sequence similarity 135, member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177819.3 | Intron | NP_808487.2 |