Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGTGGTTTCCAGGAGCTCCACCT[A/C]CAGGACTGGGATTTCCCTCCACGCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sh3d19 PubMed Links |
Sh3d19 - SH3 domain protein D19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082414.2 | Intron | NP_001075883.2 |