Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCTTTAAATAAGACACCAGGTC[C/G]TGTTTTCACTTTTTAAAATATGTGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Grhl2 PubMed Links |
Grhl2 - grainyhead-like 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026496.4 | Intron | NP_080772.2 |