Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATTTTCCTGGGAACTTGATGTG[G/T]CTTTTTTTTTTAAGTTATGTTTCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cacna2d1 PubMed Links |
Cacna2d1 - calcium channel, voltage-dependent, alpha2/delta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110843.1 | Intron | NP_001104313.1 | ||||
NM_001110844.1 | Intron | NP_001104314.1 | ||||
NM_001110845.1 | Intron | NP_001104315.1 | ||||
NM_001110846.1 | Intron | NP_001104316.1 | ||||
NM_009784.2 | Intron | NP_033914.1 |