Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAGAATCAGGCCAAAAATTGCAT[A/C]ATGAAGCATAGCTCATCAAATTCCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sgk1 PubMed Links |
Sgk1 - serum/glucocorticoid regulated kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161845.2 | Intron | NP_001155317.2 | ||||
NM_001161847.2 | Intron | NP_001155319.1 | ||||
NM_001161848.2 | Intron | NP_001155320.1 | ||||
NM_001161849.2 | Intron | NP_001155321.1 | ||||
NM_001161850.2 | Intron | NP_001155322.1 | ||||
NM_011361.3 | Intron | NP_035491.1 |