Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bmp2 PubMed Links |
Bmp2 - bone morphogenetic protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007553.3 | Intron | NP_031579.2 |
Mir3090 - microRNA 3090 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |