Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAGGAAACATTGAATCCCTGC[C/T]TAAAAGTTTCAACACTGTGAAATCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1110008P14Rik PubMed Links |
1110008P14Rik - RIKEN cDNA 1110008P14 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Ciz1 - CDKN1A interacting zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252534.1 | Intron | NP_001239463.1 | ||||
NM_001252536.1 | Intron | NP_001239465.1 | ||||
NM_001252537.1 | Intron | NP_001239466.1 | ||||
NM_001252538.1 | Intron | NP_001239467.1 | ||||
NM_028412.2 | Intron | NP_082688.1 |