Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTTCCAATTAAGAATTGGATGG[C/A]TGGCTGAAGTTTGAGGTCATCCCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mthfd1l PubMed Links |
Mthfd1l - methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170785.1 | Intron | NP_001164256.1 | ||||
NM_001170786.1 | Intron | NP_001164257.1 | ||||
NM_172308.4 | Intron | NP_758512.3 |