Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATATTACACACTTGTTTCGATGC[T/C]ATCAAATGTATTTTCTATGGGCATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Helz PubMed Links |
Helz - helicase with zinc finger domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198298.1 | Intron | NP_938040.1 |