Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGGTTTAGTGAAGAACCACAG[A/G]GGTCACTTCCCCCACTGTGTTTGGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Plch1 PubMed Links |
Plch1 - phospholipase C, eta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177732.1 | Intron | NP_001171203.1 | ||||
NM_001177733.1 | Intron | NP_001171204.1 | ||||
NM_183191.3 | Intron | NP_899014.2 |