Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTCTGGACAGGTCCGGGAGGC[A/G]GATCTGAGCGACCAGTACGAGGCAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930405A21Rik PubMed Links |
4930405A21Rik - RIKEN cDNA 4930405A21 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Dlgap4 - discs, large homolog-associated protein 4 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042487.1 | 1839 | Intron | NP_001035952.1 | |||
NM_001042488.2 | 1839 | Intron | NP_001035953.1 | |||
NM_001277186.1 | 1839 | Silent Mutation | NP_001264115.1 | |||
NM_001277187.1 | 1839 | Silent Mutation | NP_001264116.1 | |||
NM_146128.6 | 1839 | Silent Mutation | NP_666240.4 |