Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCAGAACACGGGGATGCCTCCA[A/G]GGAGCTGTTCAGGTACTCCTGTTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hs1bp3 PubMed Links |
Hs1bp3 - HCLS1 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021429.3 | 953 | Missense Mutation | NP_067404.2 |