Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATACCCTTTTGTTTTATTCTACTC[G/T]AAGTTCCATGGGCTTGAAATGTGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kmt2e PubMed Links |
Kmt2e - lysine (K)-specific methyltransferase 2E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026984.1 | 1627 | Silent Mutation | NP_081260.1 |