Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTAAGAGAAAATGTTTGTGTA[C/T]TGCTCCAAATCATGTGCTTCTTGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gata6 PubMed Links |
Gata6 - GATA binding protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010258.3 | 2389 | UTR 3 | NP_034388.2 |