Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCTTCCTACTCACTGTAGATA[C/T]TTACATTTTAACTGGCTGATGCCCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sema3c PubMed Links |
Sema3c - sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013657.5 | Intron | NP_038685.3 |